missing translation for 'onlineSavingsMsg'
Learn More

MANSC1 Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 25586-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16824145

  • 1236.20 SEK / 20µL
Estimated Shipment: 02-12-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

MANSC1
0.36 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9H8J5
9130403P13Rik, LOH12CR3, MANSC domain containing 1, MANSC1
Rabbit
IgG
20 μL
-20°C
Primary
54682
Western Blot
Unconjugated
MANSC1
Liquid
MANSC1
MANSC1 Fusion Protein Ag22357
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Videos
SDS
Documents
Certificates
Special Offers

Special Offers