Läs mer
Maolate, TRC
Produktkod. 30138630
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138630 1 g
30134097 500 mg
30011066 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30138630 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM8867401G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 886-74-8
Molekylformel C10 H12 Cl N O4
Molekylvikt (g/mol) 245.66
Synonym Chlorphenesin Carbamate, 1,2-Propanediol, 3-(4-chlorophenoxy)-, 1-carbamate, 1,2-Propanediol, 3-(p-chlorophenoxy)-, 1-carbamate (8CI), Carbamic acid, 3-(p-chlorophenoxy)-2-hydroxypropyl ester (7CI), 3-(4-Chlorophenoxy)-2-hydroxypropyl carbamate, 3-(p-Chlorophenoxy)-2-hydroxypropyl carbamate, 3-[p-Chlorophenoxy]-1,2-propanediol-1-carbamate, Chlorphenesin carbamate, Maolate, NSC 82943, Rhnesicn, Rinlaxer, U 19646, U-19,646
IUPAC-namn [3-(4-chlorophenoxy)-2-hydroxypropyl] carbamate
LEDER NC(=O)OCC(O)COc1ccc(Cl)cc1
Specifikationer
Kemiskt namn eller material Chlorphenesin Carbamate
Kvantitet 1 g
InChI formel InChI=1S/C10H12ClNO4/c11-7-1-3-9(4-2-7)15-5-8(13)6-16-10(12)14/h1-4,8,13H,5-6H2,(H2,12,14)
Formel vikt 245.0455