missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAP2K6 (MKK6) Recombinant Human Protein
MAP2K6 (MKK6) Recombinant Human Protein
Brand: Invitrogen PV3318
Additional Details : Weight : 3.36000kg
Description
MAP2K6 (MKK6) is a serine⁄threonine kinase that functions as an activator of p38 in the MAPK pathway.Specifications
NP_002749 | |
Cell Viability, Pharma and Biopharma, Cell Proliferation and Function, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
41.6kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
MAP2K6 (MKK6) | |
MAP2K6 (MKK6) was pre-diluted in enzyme dilution buffer and assayed in 24mM Tris (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 2.4mM DTT, 0.012% Triton™ X-100, 60mM NaCl, 20% glycerol, 25μM ATP, 200μg/mL inactive MAPK14 (PV3305) and trace [32P]-γ-ATP for 10 minutes at 37°C | |
MAP2K6 (MKK6) Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.41mg/mL | |
5608 | |
MAP2K6 (MKK6) Recombinant Human Protein | |
–80°C | |
1 Each | |
MAP2K6 (MKK6) Recombinant Human Protein | |
STE7 | |
9.8nmoles of phosphate transferred to inactive MAPK14 (PV3305) per minute per mg of total protein at 37°C. Activity determined at a final protein concentration of 10μg/mL | |
His Tag | |
Kinase and inhibitors | |
90% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.