missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MAPK1 (ERK2), GST Tag Recombinant Protein
Human MAPK1 (ERK2), GST Tag Recombinant Protein
Brand: Invitrogen PV3595
Additional Details : Weight : 0.05250kg
Description
MAPK1 (ERK2) is a serine⁄threonine kinase that acts as an important activator of p90 RSK, MSK, ELK1, and Stat3, and is involved in the MAPK pathway.Specifications
NP_620407 | |
69.7 kDa | |
MAPK1 (ERK2) | |
Human | |
GST Tag | |
Ser⁄Thr Kinases: CMGC Group | |
E. coli | |
Recombinant, Full Length | |
Serine/Threonine Kinases |
Kinase Assay | |
1mg | |
MAP Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid | |
Signal Transduction |
For Research Use Only. Not for use in diagnostic procedures.