missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MAPK12 (p38 γ) Recombinant Human Protein

MAPK12 (p38 gamma) is a serine/threonine kinase involved in many different cellular processes.

Brand:  Invitrogen™ PR6830A

Additional Details : Weight : 0.01000kg

Product Code. 15428493

  • 15570.00 SEK / 100µg
Estimated Shipment: 02-02-2022

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MAPK12 (p38 gamma) is activated during hypoxia and gamma-irradiation-induced G(2) arrest.

Specifications

Specifications

NP_002960
Kinase Assay
Frozen
46kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
MAP Kinase Family
Yes
Baculovirus
Protein Biology, Enzyme & Protein Activity Assays, Kinase Activity, Protein Assays and Analysis
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
100μg
RUO - research use only
MAPK12 (p38 γ)
Human
His Tag
Videos
SDS
Documents
Certificates
Special Offers

Special Offers