missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAPK4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18738-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Mitogen-activated protein kinase 4 is a member of the mitogen-activated protein kinase family. Tyrosine kinase growth factor receptors activate mitogen-activated protein kinases which then translocate into the nucleus where it phosphorylates nuclear targets.Specifications
MAPK4 | |
Polyclonal | |
Unconjugated | |
MAPK4 | |
ERK 4, ERK3, Erk4, MAP kinase 4, MAP kinase isoform p63, MAPK 4, MAPK4, p63 MAPK, p63MAPK, PRKM4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
225724, 54268, 5596 | |
-20°C | |
Liquid |
Western Blot | |
0.22 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P31152, Q63454, Q6P5G0 | |
Mapk4 | |
Peptide | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |