Learn More
Invitrogen™ MAPKAPK2 Recombinant Human Protein
MAPKAPK2 mediates p38 and ERK signaling in vivo.
Brand: Invitrogen™ PR5320A
Additional Details : Weight : 0.01000kg
Description
MAPKAPK2 is known to be involved in many cellular processes including stress and inflammatory responses, nuclear export, gene expression regulation, and cell proliferation.
Specifications
NP_116584 | |
50mM Tris, pH 7.5, 150mM NaCl, 0.05mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
MAPKAPK2, MK2 | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
48.8kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
MAPKAPK Kinase Family | |
Recombinant | |
E. coli |