missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ MAPRE2 Monoclonal Antibody (KT52)
GREENER_CHOICE

Produktkod. 17896448
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17896448 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 17896448 Leverantör Invitrogen™ Leverantörsnummer MA547664

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

Rat Monoclonal Antibody

The antibody detects a 36 kDa* protein corresponding to the molecular mass of EB2 on SDS-PAGE immunoblots of human HeLa, mouse brain, and rat PC12 cells. The antibody also detects EB2 by immunocytochemistry in HeLa and A431 cells.

The protein encoded by this gene shares significant homology to the adenomatous polyposis coli (APC) protein-binding EB1 gene family. The function of this protein is unknown; however, its homology suggests involvement in tumorigenesis of colorectal cancers and proliferative control of normal cells. This gene may belong to the intermediate/early gene family, involved in the signal transduction cascade downstream of the TCR. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen MAPRE2
Användningsområden Western Blot, Immunocytochemistry
Klassificering Monoclonal
Klona KT52
Koncentration ~1.0 mg/mL
Konjugera Unconjugated
Formulering PBS with 0.05% sodium azide
Gen MAPRE2
Genaccessionsnr. Q15555, Q3B8Q0, Q8R001
Gene Alias AI314113; APC-binding protein EB1; APC-binding protein EB2; C820009F03Rik; CSCSC2; D18Abb1e; EB1; EB2; End-binding protein 2; Mapre2; microtubule associated protein RP/EB family member 2; microtubule-associated protein RP/EB family member 2; microtubule-associated protein, RP/EB family, member 2; RP1; T-cell activation protein, EB1 family; T-cell activation protein, EB1 family, member 2
Gensymboler MAPRE2
Värd art Rat
Immunogen Clone KT52 was generated from a GST fusion protein containing human EB2. This sequence is highly conserved in rat and mouse EB2.
Reningsmetod Protein G
Kvantitet 100 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 10982, 212307, 679221
Målarter Human, Mouse, Rat
Innehåll och lagring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotyp IgG2b
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.