missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ MARCH10 Polyclonal Antibody
Goat Polyclonal Antibody
Brand: Invitrogen™ PA518853
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
This antibody is predicted to react with bovine and human based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
MARCH10 is a member of the MARCH family of membrane-bound E3 ubiquitin ligases to target lysines in substrate proteins, thereby signaling their vesicular transport between membrane compartments.
Specifikationer
| MARCH10 | |
| Polyclonal | |
| Unconjugated | |
| MARCH10 | |
| 10-Mar; 4933417C16Rik; AV039223; FLJ35757; March10; MARCH-X; membrane associated ring finger 10; membrane associated ring-CH-type finger 10; membrane-associated ring finger (C3HC4) 10; membrane-associated ring finger (C3HC4) 10, E3 ubiquitin protein ligase; membrane-associated RING finger protein 10; Membrane-associated RING-CH protein X; microtubule-associated E3 ubiquitin ligase; OTTMUSG00000002847; Probable E3 ubiquitin-protein ligase MARCH10; RGD1311692; RING finger protein 190; RING-type E3 ubiquitin transferase MARCH10; Rnf190; testis secretory sperm-binding protein Li 228n | |
| Goat | |
| Ammonium Sulfate Precipitation | |
| RUO | |
| 162333, 303596 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q5XIV2, Q8NA82 | |
| MARCH10 | |
| Peptide with sequence C-SRNYPQPRTEE, from the internal region of the protein sequence (aa 778-788) (aa 778-788). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering