Läs mer
Maritoclax, MedChemExpress
Produktkod. 16456836
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
100mg
10mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16456836 100 mg 100mg
16442767 50 mg -
16436688 10 mg 10mg
16409247 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16456836 Leverantör Medchem Express Leverantörsnummer HY15613
MedChemExpress Maritoclax (Marinopyrrole A) is a novel and specific Mcl-1 inhibitor with an IC50 value of 10.1 μM, and shows >8 fold selectivity than BCL-xl (IC50 > 80 μM).
Maritoclax (Marinopyrrole A) is a novel and specific Mcl-1 inhibitor with an IC50 value of 10.1 μM, and shows >8 fold selectivity than BCL-xl (IC50 > 80 μM).
Specifikationer
Kemiskt namn eller material Maritoclax
CAS 1227962-62-0
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C22H12Cl4N2O4
Kvantitet 100 mg
Synonym Marinopyrrole A
Löslighetsinformation DMSO : ≥ 43 mg/mL (84.29 mM)
LEDER OC1=CC=CC=C1C(C2=C(N3C(C(C4=CC=CC=C4O)=O)=CC(Cl)=C3Cl)C(Cl)=C(Cl)N2)=O
Molekylvikt (g/mol) 510.15
Formel vikt 510.15
Procent renhet 95.16%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-Kinase/protease