Läs mer
(±)-Massoilactone, TRC
Produktkod. 30471888
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30471888 1 g
1 options
Produktkod. 30471888 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM1976001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material (±)-Massoilactone
CAS 54814-64-1
Molekylformel C10H16O2
Kvantitet 1 g
Synonym 5,6-Dihydro-6-pentyl-2H-pyran-2-one, 5-Hydroxy-2-decenoic Acid Δ-Lactone, (±)-Massoia Lactone, (±)-Massoja Decalactone, 2-Decen-5-olide
InChI formel InChI=1S/C10H16O2/c1-2-3-4-6-9-7-5-8-10(11)12-9/h5,8-9H,2-4,6-7H2,1H3
LEDER O=C1OC(CCCCC)CC=C1
IUPAC-namn 6-pentyl-5,6-dihydro-2H-pyran-2-one
Molekylvikt (g/mol) 168.23
Formel vikt 168.12