Läs mer
Thermo Scientific™ Mast Cell Degranulating Peptide HR-1
Kvantitet:
1 mg
5 mg
Förpackningsstorlek:
1mg
5mg
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifikationer
Specifikationer
| Reningsmetod | HPLC |
| Formatera | Powder |
| För användning med (applikation) | Degranulates mast cells, initiating histamine release |
| Molekylvikt (g/mol) | 1492.96 |
| Produkttyp | Mast Cell Degranulating Peptide HR-1 |
| Kvantitet | 1 mg |
| Regulatorisk status | RUO - research use only |
| Sekvens | Ile-Asn-Leu-Lys-Ala-Ile-Ala-Ala-Leu-Val-Lys-Lys-Val-Leu-NH2 |
For Research Use Only