Learn More
Masterflex™ Tygon™ Fuel & Lubricant I/P™ Precision Tubing
Use this tubing for applications involving fuels and industrial lubricants. Masterfelx™ Tygon™ Fuel & Lubricant I/P™ Precision Tubing is deal for transferring gasoline, kerosene, heating oils, cutting compounds and glycol-based coolants.
7045.00 SEK - 8015.00 SEK
Specifications
Product Type | Pump Tubing |
---|---|
Material | Thermoplastic |
Overall Length (Metric) | 1500 cm |
For Use With (Application) | transferring Hydrocarbons, Gasoline, Kerosene, heating oils, cutting compounds, and Glycol based coolants |
Min. Operating Temperature (Metric) | -37°C |
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
11795099
|
Masterflex™
06401-73 |
73 |
7045.00 SEK
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||||
11730409
|
Masterflex™
06401-82 |
82 |
8015.00 SEK
Each |
Estimated Shipment: 01-07-2024
Log in to see stock. |
|||||
Description
- Resists most hydrocarbons, embrittlement and swelling
- Very low gas permeability
- Complies with USP Class V, EP, FDA, REACH, RoHS, ADCF
- Ozone- and UV-resistant
- Low extractability
- Translucent yellow High dielectric constant
- Thermoplastic
- PVC-based material with plasticizer
Specifications
Pump Tubing | |
1500 cm | |
-37°C | |
Low | |
74°C |
Thermoplastic | |
transferring Hydrocarbons, Gasoline, Kerosene, heating oils, cutting compounds, and Glycol based coolants | |
1 Each | |
Translucent Yellow |