Läs mer
Ismatec™ Masterflex™ PharMed™ Ismaprene Standard Tubing
Slangstorlek:
1.6mm I.D., 4.8mm O.D.
4.8mm I.D., 8.0mm O.D.
6.4mm I.D., 12.8mm O.D.
9.5mm I.D., 15.9mm O.D.
Förpackningsstorlek:
7.5m
Beskrivning
- Ideal for production filtration, fermentation and bioreactor process lines.
- Very long service-life.
- Non-toxic and non-hemolytic.
- Impermeable to normal light and UV-radiation
- Appropriate for medical products and foodstuffs.
- Low particulate spallation.
- Can be autoclaved repeatedly.
- Withstands repeated CIP and SIP cleaning and sterilization.
- Meets USP class VI, FDA and NSF criteria.
- Potential leaching of additives (lubricants).
- Applications
- Acids: Good
- Alkaline solutions: Good
- Solvents: Not recommended
- Pressure: Not recommended
- Viscous media: Good
- Sterile media: Excellent
- Complies with: FDA 21CFR177.2600, US Pharmacopoea Class VI, NSF listed (Standard 51).
- Sterilization
- Autoclavable with steam, 30 minutes at 1 bar (15 psi) and 141°C.
- Gas sterilization with ethylene oxide.
- Sterilization with radiation up to 2.5 mrad.
- Caution: Use special tubing version (welded stoppers) when autoclaving 2 or 3-stop color-coded tubing.
Specifikationer
Specifikationer
| Certifikat och efterlevnad | FDA 21CFR177.2600, US Pharmacopoea Class VI, NSF listed (Standard 51) |
| Autoklaveringsbar | Autoclavable |
| Material | PharMed Ismaprene |
| Total längd (metrisk) | 7.5 m |
| Tjocklek (metrisk) | 3.2 mm |
| För användning med (applikation) | Pharmaceutical, medical, foodstuffs |
| Min. Driftstemperatur (metrisk) | -60°C |
| Slangstorlek | 9.5mm I.D., 15.9mm O.D. |
| Gaspermeabilitet | CO2 1200, 02 200, N2 80 |
| Färg | Beige |
| Visa mer |