Learn More
Masterflex™ Polypropylene Male Luer Adapters
Brand:  Masterflex™ 45518-11
Additional Details : Weight : 0.90000kg
Product Code. 15298744
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
Luer Adapter
Fits Male Luer Lock to 1/18 in. I.D. low profile barb
25 Pack
Polypropylene
Ideal for a variety of applications, including syringes or small tubing