Learn More
Masterflex™ Single-channel Series Q Pump Head Drive Adapter Kit
Product Code. 11755848 Shop All Masterflex Products
Change view
Click to view available options
No. of Channels:
1
Unit Size:
Each
Product Code. No. of Channels Unit Size
11755848 1 Each
1 options
Product Code. 11755848 Supplier Masterflex™ Supplier No. 0710448
  • ONLINE EXCLUSIVE PRICE
    14190.00 SEK
    10742.70 SEK / Paket med 1
    Save 3447.30 SEK
    24% Av
Beräknad leverans: 23-02-2027
Specifications
For Use With (Application) To attach Micropump™ A-Mount pump heads to your own motor or Masterflex™ L/S™ pump drives
No. of Channels 1
Quantity 1 Set
Type Series Q Pump Head Drive Adapter Kit