Läs mer
Masterflex™ I/P™ Brushless Process Drive with High-Performance Pump Head for High-Performance Precision Tubing
Flödeshastighet:
17 l/min
Förpackningsstorlek:
Varje
Beskrivning
- Digital interface with LED display shows percent speed with 1% resolution
- Speed control via three-turn potentiometer for quick, easy adjustments
- Easily control start/stop and reverse features via sealed keypad
- Constructed of non-chip painted steel and aluminum IP55-rated housing
Specifikationer
Specifikationer
| Certifikat och efterlevnad | CE, cULus, ISO 9001:2008 |
| Kontrolltyp | Variable Speed |
| Visningstyp | LED |
| Kapslingsklassificering | IP55 |
| Flödeshastighet | 17 l/min |
| Motorhastighet | 650 rpm |
| Avsluta | Epoxy-Coated |
| Hästkraft | 1/3 hp |
| Inkluderar | 06429-88, 3 ft. (1m) Tubing, 77410-10 Drive, I/P High-Performance 77600-62 Pump Head, Tygon LFLI/P88 |
| Material | Aluminum, Steel |
| Visa mer |