missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MASTL, GST Tag Recombinant Protein
This MASTL Recombinant Human Protein (full length) was expressed in insect cells.
4940.00 SEK - 72900.00 SEK
Specifications
Accession Number | NP_001165774 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 123.6 kDa |
KinaseFamily | MAST Family |
Protein Length | Full Length |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15960303
|
Invitrogen™
A30978 |
10 μg |
4940.00 SEK
10µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15980383
|
Invitrogen™
A32896 |
100 μg |
23360.00 SEK
100µg |
Estimated Shipment: 10-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17158795
|
Invitrogen™
A33381 |
1 mg |
72900.00 SEK
1mg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MASTL or microtubule associated serine/threonine kinase-like belongs to the MAST family, AGC Ser/Thr protein kinase group. MASTL promotes mitotic progression and cell cycle reentry after DNA damage. Loss of MASTL leads to defects in chromosome condensation, separation, and other aspects of mitotic progression, which is primarily mediated by inhibiting PP2A/B55 delta. Overexpression of MASTL is correlated with cancer progression, poor patient survival, and tumor recurrence in various cancers.- Validated for use with the LanthaScreen™ Eu Kinase Binding assay
- Available in multiple pack sizes for convenient use
Specifications
NP_001165774 | |
123.6 kDa | |
Full Length | |
Recombinant | |
Storage Buffer: 50 mM Tris-HCl, pH 7.5, 150 mM NaCl, 10 mM glutathione, 0.1 mM EDTA, 0.25 mM DTT, 0.1 mM PMSF, 25% glycerol. | |
Liquid |
Kinase Assay | |
MAST Family | |
Human | |
GST Tag | |
Dry Ice |