Learn More
MAT1A Mouse anti-Human, Unconjugated, Clone: 2F1C1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67408-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16870016
  • 4721.67 SEK / 150µL
Estimated Shipment: 20-05-2024
to see stock.
Description
Description
The MAT1A antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MAT1A. This antibody recognizes human, mouse, rat antigen. The MAT1A antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
MAT1A
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, ELISA
2F1C1
Unconjugated
150 μL
4143
IgG2a