Läs mer
(+)-Matrine, TRC
Produktkod. 17994915
Change view
Kvantitet:
100 mg
500 mg
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17994915 500 mg
17984455 100 mg
17971595 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17994915 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM197870500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 519-02-8
Molekylformel C15 H24 N2 O
Molekylvikt (g/mol) 248.36
Synonym 1H,5H,10H-Dipyrido[2,1-f:3',2',1'-ij][1,6]naphthyridin-10-one, dodecahydro-, (7aS,13aR,13bR,13cS)-, Matridin-15-one, Matrine (6CI,7CI,8CI), (+)-Matrine, (7aS,13aR,13bR,13cS)-Dodecahydro-1H,5H,10H-dipyrido[2,1-f:3',2',1'-ij][1,6]naphthyridin-10-one, NSC 146051, NSC 318810, Vegard, α-Matrine
LEDER O=C1CCC[C@@H]2[C@H]3CCCN4CCC[C@@H](CN12)[C@@H]34
Specifikationer
Kemiskt namn eller material Matrine
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C15H24N2O/c18-14-7-1-6-13-12-5-3-9-16-8-2-4-11(15(12)16)10-17(13)14/h11-13,15H,1-10H2/t11-,12+,13+,15-/m0/s1
Formel vikt 248.1889
Procent renhet >95%