Läs mer
Mavacoxib, TRC
Produktkod. 30137306
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137306 1 g
30006330 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30137306 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM1979001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 170569-88-7
Molekylformel C16H11F4N3O2S
Molekylvikt (g/mol) 385.34
LEDER NS(=O)(=O)c1ccc(cc1)n2nc(cc2c3ccc(F)cc3)C(F)(F)F
Specifikationer
Kemiskt namn eller material Mavacoxib
Kvantitet 1 g