missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MAX (Human) Recombinant Protein
Human MAX full-length ORF ( AAH03525, 1 a.a. - 151 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004149-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MSDNDDIEVESDADKRAHHNALERKRRDHIKDSFHSLRDSVPSLQGEKASRAQILDKATEYIQYMRRKNHTHQQDIDDLKRQNALLEQQVRALEKARSSAQLQTNYPSSDNSLYTNAKGSTISAFDGGSDSSSESEPEEPQSRKKLRMEAS
Specifications
AAH03525 | |
MYC associated factor X | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MAX | |
GST |
4149 | |
Wheat germ expression system | |
10 μg | |
MGC10775, MGC11225, MGC18164, MGC34679, MGC36767, bHLHd4, bHLHd5, bHLHd6, bHLHd7, bHLHd8, orf1 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |