Learn More
Thermo Scientific™ MaxQ™ 7000 Dedicated Platforms with Clamps
Maximize capacity when working with a single-size vessel by using these dedicated platforms with clamps.
Brand:  Thermo Scientific™ 3540-4
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 10013123
  • 6969.50 SEK / Each
Estimated Shipment: 26-11-2021
Add to basket
Description
Description
Maximize capacity when working with a single-size vessel by using Thermo Scientific™ MaxQ™ 7000 Dedicated Platforms with Clamps. With exceptional innovation, reliability and accuracy, MaxQ 7000 shakers, platforms and accessories are the optimal choice for a busy lab.
Clamps come installed to provide maximum capacity.
Requires:
Universal platform and clamps or dedicated platform with preinstalled clamps
Specifications
Specifications
16
11 in. x 13 in.
11 in.
279.4 mm
125mL Erlenmeyer Flask
MaxQ™ 7000 Shaker
Certificate of Analysis (COA), Certificate of Conformance (COC)
MaxQ 7000
13 in.
330.2 mm
Shaker Platform with Clamps