missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBIP Mouse anti-Human, Clone: 2E6H11, Proteintech
Mouse Monoclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | MBIP |
---|---|
Clone | 2E6H11 |
Concentration | 1.21 mg/mL |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16827675
|
Proteintech
66102-1-IG-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16817675
|
Proteintech
66102-1-IG-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
MBIP inhibits the MAP3K12 activity to induce the activation of the JNK/SAPK pathway. Component of the ATAC complex, a complex with histone acetyltransferase activity on histones H3 and H4.Specifications
MBIP | |
1.21 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
MBIP, MUK binding inhibitory protein | |
MBIP | |
IgG2b | |
Protein A | |
Antibody |
2E6H11 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Q9NS73 | |
51562 | |
MBIP Fusion Protein Ag18090 | |
Primary | |
-20°C | |
MBIP |