Läs mer
MBL™ Blue Spirit Total Immersion Thermometer
Produktkod. 12033823
Change view
Temperaturområde (metriskt):
-20 °C till +150 °C
Förpackningsstorlek:
Varje
Produktkod. Temperaturområde (metriskt) unitSize
12033823 -20 °C till +150 °C Varje
1 options
Produktkod. 12033823 Leverantör MBL International Leverantörsnummer 30/150/10
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Filled with safe low toxicity blue or green indicating fluid and calibrated for total immersion. MBL™ Blue Spirit Total Immersion Thermometer gives accurate readings when the thermometer is immersed to a specific depth. Graduations are marked in permanent amber stain. White background provides clear observation of colored liquid column. Reinforced bulb allows stirring. Includes plastic non-roll top and safety suspension ring.
  • Mercury free - filled with a low toxicity blue or green spirit
  • Calibrated for total immersion
  • Permanent amber stain graduations
  • White colored background provides clear observation of blue colored liquid column
  • Reinforced bulb for stirring
  • With plastic non-roll top and safety suspension ring
  • Nominal length 305mm with item 30/210/10 length 405mm
TRUSTED_SUSTAINABILITY
Specifikationer
Typ av nedsänkning Total
Inkluderar Safety suspension ring
Längd (metrisk) Totalt 305 mm
Termometerfyllning Mercury-free
Produkttyp Nonmercury Thermometer
Max. Temperatur (metrisk) 150°C
Min. Temperatur (metrisk) -20°C
Temperaturområde (metriskt) -20 °C till +150 °C