Learn More
MBL™ Class A Kohlrausch Flask
Expanded neck for easy introduction of viscous liquids or solid samples.
Brand: MBL FRE013
Additional Details : Weight : 0.27000kg
Description
Expanded neck for easy introduction of viscous liquids or solid samples.
- Complies with BS 615
- With expanded neck for easy introduction of viscous liquids or solid samples
- 2 sizes available: 100mL for sugar analysis, 200mL for determination of oil viscosity by Engler method
- Calibrated to Class A tolerances
- White enamel inscriptions
- Manufactured from soda-lime glass
Specifications
200 mL | |
185 mm | |
Kohlrausch flask |
Borosilicate Glass | |
2 |