missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
MC180295, MedChemExpress
Beskrivning
MC180295 ((rel)-MC180295) is a potent and selective CDK9-Cyclin T1 inhibitor, with an IC50 of 5 nM, at least 22-fold more selective for CDK9 over other CDKs. MC180295 also inhibits GSK-3α and GSK-3β. MC180295 ((rel)-MC180295) has potent anti-tumor effect.
Specifikationer
Specifikationer
| Kemiskt namn eller material | MC180295 |
| Fysisk form | Solid |
| CAS | 2237942-08-2 |
| Färg | Orange |
| Kvantitet | 50 mg |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C17H18N4O3S |
| För användning med (applikation) | Cancer-Kinase/protease |
| Synonym | (rel)-MC180295 |
| Löslighetsinformation | DMSO : 100 mg/mL (279.01 mM; Need ultrasonic) |
| Visa mer |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?