missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ MCEE Monoclonal Antibody (OTI6A1)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527015
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
MCEE Monoclonal Antibody for Western Blot, IHC (P)
MCEE catalyzes the interconversion of D- and L-methylmalonyl-CoA during the degradation of branched chain amino acids. odd chain-length fatty acids, and other metabolites. Mutations in this gene result in methylmalonyl-CoA epimerase deficiency, which is presented as mild to moderate methylmalonic aciduria.
Specifikationer
| MCEE | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q96PE7 | |
| MCEE | |
| Full length human recombinant protein of MCEE produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6A1 | |
| Unconjugated | |
| MCEE | |
| 1110007A04Rik; DL-methylmalonyl-CoA racemase; GLOD2; glyoxalase domain containing 2; Mcee; methylmalonyl CoA epimerase; methylmalonyl-CoA epimerase; methylmalonyl-CoA epimerase, mitochondrial | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 84693 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction