missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15597-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is one of the highly conserved mini-chromosome maintece proteins (MCM) that are involved in the initiation of eukaryotic genome replication. The hexameric protein complex formed by MCM proteins is a key component of the pre-replication complex (pre_RC) and may be involved in the formation of replication forks and in the recruitment of other DNA replication related proteins. This protein is a subunit of the protein complex that consists of MCM2-7. It has been shown to interact directly with MCM5/CDC46. This protein also interacts with, and thus is acetlyated by MCM3AP, a chromatin-associated acetyltransferase. The acetylation of this protein inhibits the initiation of DNA replication and cell cycle progression.Specifications
MCM3 | |
Polyclonal | |
Unconjugated | |
MCM3 | |
HCC5, MCM3, P1 MCM3, P1.h, p102, RLF subunit beta, RLFB | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17215, 316273, 4172 | |
-20°C | |
Liquid |
Western Blot, Immunoprecipitation | |
0.18 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P25205, P25206 | |
Mcm3 | |
MCM3 Fusion Protein Ag7966 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |