missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11703-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is structurally very similar to the CDC46 protein from S. cerevisiae, a protein involved in the initiation of DNA replication. The encoded protein is a member of the MCM family of chromatin-binding proteins and can interact with at least two other members of this family. The encoded protein is upregulated in the transition from the G0 to G1/S phase of the cell cycle and may actively participate in cell cycle regulation.Specifications
MCM5 | |
Polyclonal | |
Unconjugated | |
MCM5 | |
CDC46, CDC46 homolog, MCM5, P1 CDC46 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17218, 291885, 4174 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P33992, P49718 | |
Mcm5 | |
MCM5 Fusion Protein Ag2271 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |