missing translation for 'onlineSavingsMsg'
Learn More

MCM7 Mouse, Clone: MCM7/1466, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 4176-MSM1-P0

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15950448

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

MCM7
Nuclear
MCM7/1466
Unconjugated
NP_005907
Mouse
88kDa
Protein A/G Purified
RUO
4176
IgG2b
Western Blot
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
MCM7
Recombinant human MCM7 protein fragment (aa195-319)
Raji or Ramos cells. Tonsil
20 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers