Läs mer
Tocris Bioscience™ McN-A 343
GREENER_CHOICE
Produktkod. 18034162
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18034162 50 mg
18004922 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18034162 Leverantör Tocris Bioscience™ Leverantörsnummer 1384/50
Selective M1 muscarinic agonist
Selective muscarinic M1 receptor agonist. Selectivity for M1 over other muscarinic receptor types appears to arise from a high efficacy at M1 receptors.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55-45-8
Molekylformel C14H18Cl2N2O2
Specifikationer
Kemiskt namn eller material McN-A 343
Kvantitet 50 mg
Formel vikt 317.21
Renhet >99%
Molekylvikt (g/mol) 317.21