Läs mer
MCOPPB Trihydrochloride, TRC
Produktkod. 30479504
Change view
Kvantitet:
50 mg
Produktkod. Kvantitet
30479504 50 mg
1 options
Produktkod. 30479504 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM19950350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material MCOPPB (hydrochloride)
CAS 1108147-88-1
Molekylformel C26H40N4 . 3(HCl)
Kvantitet 50 mg
Synonym 1-[1-(1-Methylcyclooctyl)-4-piperidinyl]-2-(3R)-3-piperidinyl-1H-benzimidazole trihydrochloride, 1-[1-(1-Methylcyclooctyl)-4-piperidinyl]-2-(3R)-3-piperidinyl-1H-benzimidazole Hydrochloride (1:3)
InChI formel InChI=1S/C26H40N4.3ClH/c1-26(15-7-3-2-4-8-16-26)29-18-13-22(14-19-29)30-24-12-6-5-11-23(24)28-25(30)21-10-9-17-27-20-21;;;/h5-6,11-12,21-22,27H,2-4,7-10,13-20H2,1H3;3*1H/t21-;;;/m1.../s1
LEDER CC1(CCCCCCC1)N2CCC(CC2)N3C4=CC=CC=C4N=C3[C@H]5CNCCC5.Cl.Cl.Cl
IUPAC-namn (R)-1-(1-(1-methylcyclooctyl)piperidin-4-yl)-2-(piperidin-3-yl)-1H-benzo[d]imidazole trihydrochloride
Molekylvikt (g/mol) 518
Formel vikt 516.27