missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCT14 Rabbit anti-Human, Mouse, Porcine, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | MCT14 |
---|---|
Concentration | 0.37 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16825285
|
Proteintech
26953-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16815285
|
Proteintech
26953-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 24-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
MCT14 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Porcine | |
Q7RTX9, Q8K1C7 | |
100519114, 151473, 71781 | |
MCT14 Fusion Protein Ag25315 | |
Primary | |
-20°C | |
SLC16A14 |
0.37 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
MCT 14, MCT14, Monocarboxylate transporter 14, SLC16A14 | |
Slc16a14 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |