Learn More
mDia1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 20624-1-AP-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16840185
  • 4721.67 SEK / 150µL
Estimated Shipment: 10-06-2024
to see stock.
Description
Description
The mDia1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human mDia1. This antibody recognizes human, mouse, rat, monkey antigen. The mDia1 antibody has been validated for the following applications: IF, IP, WB, ELISA analysis.
Specifications
Specifications
mDia1
Polyclonal
Unconjugated
150 μL
1729
IgG
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence
207 μg/mL
Rabbit
Primary
Human, Mouse, Rat, Monkey