Learn More
mDia1 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20624-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
The mDia1 antibody from Proteintech is a rabbit polyclonal antibody generated with fusion protein of human mDia1. This antibody recognizes human, mouse, rat, monkey antigen. The mDia1 antibody has been validated for the following applications: IF, IP, WB, ELISA analysis.
Specifications
mDia1 | |
Polyclonal | |
Unconjugated | |
20 μL | |
1729 | |
IgG |
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence | |
207 μg/mL | |
Rabbit | |
Primary | |
Human, Mouse, Rat, Monkey |