missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ MDM2 (Human) Recombinant Protein
Human MDM2 full-length ORF ( ENSP00000258149, 1 a.a. - 491 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00004193-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MCNTNMSVPTDGAVTTSQIPASEQETLVRPKPLLLKLLKSVGAQKDTYTMKEVLFYLGQYIMTKRLYDEKQQHIVYCSNDLLGDLFGVPSFSVKEHRKIYTMIYRNLVVVNQQESSDSGTSVSENRCHLEGGSDQKDLVQELQEEKPSSSHLVSRPSTSSRRRAISETEENSDELSGERQRKRHKSDSISLSFDESLALCVIREICCERSSSSESTGTPSNPDLDAGVSEHSGDWLDQDSVSDQFSVEFEVESLDSEDYSLSEEGQELSDEDDEVYQVTVYQAGESDTDSFEEDPEISLADYWKCTSCNEMNPPLPSHCNRCWALRENWLPEDKGKDKGEISEKAKLENSTQAEEGFDVPDCKKTIVNDSRESCVEENDDKITQASQSQESEDYSQPSTSSSIIYSSQEDVKEFEREETQDKEESVESSLPLNAIEPCVICQGRPKNGCIVHGKTGHLMACFTCAKKLKKRNKPCPVCRQPIQMIVLTYFP
Specifications
ENSP00000258149 | |
Mdm2 p53 binding protein homolog (mouse) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MDM2 | |
GST |
4193 | |
Wheat germ expression system | |
25 μg | |
HDMX, MGC71221, hdm2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |