missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
MDR3/ABCB4 Antibody (P3II-26), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21455
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
MDR3/ABCB4 Monoclonal antibody specifically detects MDR3/ABCB4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Frozen)
Specifikationer
| MDR3/ABCB4 | |
| Monoclonal | |
| Unconjugated | |
| PBS, 0.1% BSA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Frozen) | |
| P3II-26 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:20-1:50, Immunohistochemistry-Frozen 1:2 | |
| ABC21, ATP-binding cassette sub-family B member 4, ATP-binding cassette, sub-family B (MDR/TAP), member 4, EC 3.6.3, EC 3.6.3.44, MDR2, MDR3MDR2/3, multidrug resistance protein 3, multiple drug resistance 3, P glycoprotein 3/multiple drug resistance 3, PFIC-3, P-glycoprotein 3, P-glycoprotein-3/multiple drug resistance-3, PGY3GBD1 | |
| Selected after immunization with a fusion protein of Gluthathione S Transferase and a fragment of MDR3/ABCB4 (amino acids 629-692) | |
| 1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 5244 | |
| Store at 4C. | |
| IgG2b |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering