Läs mer
ME0328, MedChemExpress
Produktkod. 16475597
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
25 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16475597 5 mg
16464266 100 mg
16452858 10 mg
16432546 25 mg
16409516 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16475597 Leverantör Medchem Express Leverantörsnummer HY100225
MedChemExpress ME0328 is a potent and selective ARTD3/PARP3 inhibitor with an IC50 of 0.89±0.28 μM.
ME0328 is a potent and selective ARTD3/PARP3 inhibitor with an IC50 of 0.89±0.28 μM.
Specifikationer
Kemiskt namn eller material ME0328
Fysisk form Solid
CAS 1445251-22-8
Färg White
Kvantitet 5 mg
Anteckningar om renhetsgrad Research
Molekylformel C19H19N3O2
För användning med (applikation) Cancer-programmed cell death
Löslighetsinformation DMSO : 75 mg/mL (233.38 mM; Need ultrasonic)
LEDER O=C(N[C@H](C1=CC=CC=C1)C)CCC(NC2=C3C=CC=C2)=NC3=O
Molekylvikt (g/mol) 321.37
Formel vikt 321.37
Procent renhet 99.87%