Läs mer
Mecamylamine Hydrochloride, TRC
Produktkod. 30482872
Change view
Kvantitet:
25 mg
Produktkod. Kvantitet
30482872 25 mg
1 options
Produktkod. 30482872 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM20260025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Mecamylamine Hydrochloride
CAS 826-39-1
Molekylformel C11 H21 N . Cl H
Kvantitet 25 mg
Synonym Bicyclo[2.2.1]heptan-2-amine, N,2,3,3-tetramethyl-, hydrochloride (1:1), 2-Norbornanamine, N,2,3,3-tetramethyl-, hydrochloride (8CI), Bicyclo[2.2.1]heptan-2-amine, N,2,3,3-tetramethyl-, hydrochloride (9CI), CPDD 0059, Inversine, Mecamylamine chloride, Mecamylamine hydrochloride, Mevasin, Mevasine, N,2,3,3-Tetramethyl-2-norbornanamine hydrochloride
InChI formel InChI=1S/C11H21N.ClH/c1-10(2)8-5-6-9(7-8)11(10,3)12-4;/h8-9,12H,5-7H2,1-4H3;1H
LEDER Cl.CNC1(C)C2CCC(C2)C1(C)C
IUPAC-namn N,2,2,3-tetramethylbicyclo[2.2.1]heptan-3-amine;hydrochloride
Molekylvikt (g/mol) 203.75
Formel vikt 203.1441