Läs mer
Mecillinam, 90+%, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J66774.MC
1487.80 SEK valid until 2026-07-03
BÄSTA PRIS på kampanj! Använd kampanjkod: "27965" för att få ditt kampanjpris.
| Kvantitet | 100mg |
|---|
Beskrivning
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifikationer
| Mecillinam | |
| White | |
| >110°C (230°F) | |
| MFCD00056869 | |
| 14,389 | |
| BWWVAEOLVKTZFQ-NTZNESFSSA-N | |
| (2S,5R,6R)-6-(azepan-1-ylmethylideneamino)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid | |
| 36273 | |
| 325.43 |
| 32887-01-7 | |
| 100mg | |
| C15H23N3O3S | |
| 551°C | |
| Soluble in water at approximately 1mg/mL | |
| CC1(C(N2C(S1)C(C2=O)N=CN3CCCCCC3)C(=O)O)C | |
| 325.427 | |
| CHEBI:51208 | |
| 95% |