missing translation for 'onlineSavingsMsg'
Läs mer

Meclizine Dihydrochloride, TRC

Produktkod. 30480412
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30480412 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30480412 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM20269550MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material Meclozine Hydrochloride
CAS 1104-22-9
Molekylformel C25 H27 Cl N2 . 2 Cl H
Kvantitet 50 mg
Synonym Meclozine Dihydrochloride, 1-[(4-Chlorophenyl)phenylmethyl]-4-[(3-methylphenyl)methyl]piperazine dihydrochloride, 1-(p-Chloro-α-phenylbenzyl)-4-(m-methylbenzyl)piperazine dihydrochloride, Ancolan dihydrochloride, Bonamine, Diadril, Meclizine dihydrochloride, Meclizine hydrochloride, Meclozine dihydrochloride, Meclozine hydrochloride, NSC 28728, Navidoxine, Parachloramine dihydrochloride, Postafen, Postafene dihydrochloride
InChI formel InChI=1S/C25H27ClN2.2ClH/c1-20-6-5-7-21(18-20)19-27-14-16-28(17-15-27)25(22-8-3-2-4-9-22)23-10-12-24(26)13-11-23;;/h2-13,18,25H,14-17,19H2,1H3;2*1H
LEDER Cl.Cl.Cc1cccc(CN2CCN(CC2)C(c3ccccc3)c4ccc(Cl)cc4)c1
IUPAC-namn 1-[(4-chlorophenyl)-phenylmethyl]-4-[(3-methylphenyl)methyl]piperazine;dihydrochloride
Molekylvikt (g/mol) 463.87
Formel vikt 462.1396
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.