Läs mer
Meclocycline Sulfosalicylate Salt, TRC
Produktkod. 30476369
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30476369 100 mg
1 options
Produktkod. 30476369 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM202720100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Meclocycline Sulfosalicylate
CAS 73816-42-9
Molekylformel C22 H21 Cl N2 O8 . C7 H6 O6 S
Kvantitet 100 mg
Synonym Benzoic acid, 2-hydroxy-5-sulfo-, compd. with (4S,4aR,5S,5aR,12aS)-7-chloro-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methylene-1,11-dioxo-2-naphthacenecarboxamide (1:1), Benzoic acid, 2-hydroxy-5-sulfo-, compd. with [4S-(4α,4aα,5α,5aα,12aα)]-7-chloro-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methylene-1,11-dioxo-2-naphthacenecarboxamide (1:1), Salicylic acid, 5-sulfo-, compd. with 7-chloro-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methylene-1,11-dioxo-2-naphthacenecarboxamide (7CI), 2-Naphthacenecarboxamide, 7-chloro-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methylene-1,11-dioxo-, (4S,4aR,5S,5aR,12aS)-, mono(2-hydroxy-5-sulfobenzoate) (salt) (9CI), 2-Naphthacenecarboxamide, 7-chloro-4-(dimethylamino)-1,4,4a,5,5a,6,11,12a-octahydro-3,5,10,12,12a-pentahydroxy-6-methylene-1,11-dioxo-, [4S-(4α,4aα,5α,5aα,12aα)]-, mono(2-hydroxy-5-sulfobenzoate) (salt), Meclan, Meclan Cream, Meclocycline 5-sulfosalicylate, Meclocycline sulfosalicylate, Mecloderm, Meclosorb, Meclutin, Traumatociclina
InChI formel InChI=1S/C22H21ClN2O8.C7H6O6S/c1-6-9-7(23)4-5-8(26)11(9)16(27)12-10(6)17(28)14-15(25(2)3)18(29)13(21(24)32)20(31)22(14,33)19(12)30;8-6-2-1-4(14(11,12)13)3-5(6)7(9)10/h4-5,10,14-15,17,26,28-30,33H,1H2,2-3H3,(H2,24,32);1-3,8H,(H,9,10)(H,11,12,13)/t10-,14-,15+,17+,22+;/m1./s1
LEDER CN(C)[C@H]1[C@@H]2[C@@H](O)[C@@H]3C(=C)c4c(Cl)ccc(O)c4C(=O)C3=C(O)[C@]2(O)C(=O)C(=C1O)C(=O)N.OC(=O)c5cc(ccc5O)S(=O)(=O)O
IUPAC-namn (4S,4aR,5S,5aR,12aS)-7-chloro-4-(dimethylamino)-3,5,10,12,12a-pentahydroxy-6-methylidene-1,11-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;2-hydroxy-5-sulfobenzoic acid
Molekylvikt (g/mol) 695.05
Formel vikt 694.0872