Läs mer
Meclofenamate Sodium, TRC
Produktkod. 17904015
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17904015 100 mg
17972185 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17904015 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM202740100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6385-02-0
Molekylformel C14 H10 Cl2 N O2 . Na
Molekylvikt (g/mol) 318.13
Synonym Benzoic acid, 2-[(2,6-dichloro-3-methylphenyl)amino]-, sodium salt (1:1), Anthranilic acid, N-(2,6-dichloro-m-tolyl)-, monosodium salt (8CI), Benzoic acid, 2-[(2,6-dichloro-3-methylphenyl)amino]-, monosodium salt (9CI), CI 583, Meclofenamate Sodium, Meclomen, N-(2,6-Dichloro-m-tolyl)anthranilic acid sodium salt, Sodium N-(2,6-dichloro-m-tolyl)anthranilate, Sodium meclofenamate, Sodium meclophenamate
IUPAC-namn sodium;2-(2,6-dichloro-3-methylanilino)benzoate
LEDER [Na+].Cc1ccc(Cl)c(Nc2ccccc2C(=O)[O-])c1Cl
Specifikationer
Kemiskt namn eller material Meclofenamate Sodium
Kvantitet 100 mg
InChI formel InChI=1S/C14H11Cl2NO2.Na/c1-8-6-7-10(15)13(12(8)16)17-11-5-3-2-4-9(11)14(18)19;/h2-7,17H,1H3,(H,18,19);/q;+1/p-1
Formel vikt 316.9986