Läs mer
Mecobalamin, TRC
Specifikationer
Specifikationer
| Kemiskt namn eller material | Methylcobalamin |
| CAS | 13422-55-4 |
| Anteckningar om renhetsgrad | HPLC |
| Molekylformel | C63 H91 Co N13 O14 P |
| Kvantitet | 2.5 g |
| Synonym | Cobinamide, Co-methyl-, dihydrogen phosphate (ester), inner salt, 3'-ester with (5,6-dimethyl-1-α-D-ribofuranosyl-1H-benzimidazole-κN3) (9CI), Cobinamide, Co-methyl deriv., hydroxide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-α-D-ribofuranosyl-1H-benzimidazole, Cobinamide, Co-methyl deriv., hydroxide, dihydrogen phosphate (ester), inner salt, 3'-ester with 5,6-dimethyl-1-α-D-ribofuranosylbenzimidazole (8CI), Algobaz, Co-Methylcobalamin, Cobalamin, Co-methyl-, Hitocobamin M, MeCbl, Mecobalamin, Methycobal, Methyl cobalamine, Methyl vitamin B12, Methyl-5,6-dimethylbenzimidazolylcobalamin, Methyl-B12, Methylcob(III)alamin, Methylcobalamin, Methylcobaz |
| InChI formel | InChI=1S/C62H90N13O14P.CH3.Co/c1-29-20-39-40(21-30(29)2)75(28-70-39)57-52(84)53(41(27-76)87-57)89-90(85,86)88-31(3)26-69-49(83)18-19-59(8)37(22-46(66)80)56-62(11)61(10,25-48(68)82)36(14-17-45(65)79)51(74-62)33(5)55-60(9,24-47(67)81)34(12-15-43(63)77)38(71-55)23-42-58(6,7)35(13-16-44(64)78)50(72-42)32(4)54(59)73-56;;/h20-21,23,28,31,34-37,41,52-53,56-57,76,84H,12-19,22,24-27H2,1-11H3,(H15,63,64,65,66,67,68,69,71,72,73,74,77,78,79,80,81,82,83,85,86);1H3;/q;;+2/p-2/t31-,34-,35-,36-,37+,41-,52-,53-,56-,57+,59-,60+,61+,62+;;/m1../s1 |
| LEDER | C[C@@H]1CNC(=O)CC[C@]2(C)[C@@H](CC(=O)N)[C@H]3N4C2=C(C)C5=N6C(=CC7=N8C(=C(C)C9=N([C@]3(C)[C@@](C)(CC(=O)N)[C@@H]9CCC(=O)N)[Co+]468(C)N%10=CN([C@H]%11O[C@H](CO)[C@@H](OP(=O)([O-])O1)[C@H]%11O)c%12cc(C)c(C)cc%10%12)[C@@](C)(CC(=O)N)[C@@H]7CCC(=O)N)C(C)(C)[C@@H]5CCC(=O)N |
| Molekylvikt (g/mol) | 1344.38 |
| Formel vikt | 1343.5878 |
| Visa mer |