Läs mer
AMPri™ Med-Comfort Premium Grip Latex Examination Gloves, PPE Cat. III
Beskrivning
- AQL 1.5
- Premium grip examination gloves
- Applications include laboratory, clinical, pharmaceutical, dental, and food industry use
- Protect against chemicals and microorganisms
- Ambidextrous fit
- Powder-free
- Non-sterile
- Color: white
- Textured finish
- Disposable
Specifikationer
Specifikationer
| Certifikat och efterlevnad | AQL 1.5 |
| Certifieringar/efterlevnad (Europa) | EN 455-1, EN374-1/4/5, EN420, Food grade |
| Färg | White |
| Disponibel | Single-use |
| Användning av laboratorium/renrum | Laboratory |
| Latexfri | No |
| Material | Latex |
| Storlek | Medium |
| Tjocklek (metrisk) | 0.08 mm (Cuff) |
| Längd (metrisk) | 240 mm |