missing translation for 'onlineSavingsMsg'
Learn More

MED18 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15352-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16894494

  • 1656.73 SEK / 20µL
Estimated Shipment: 17-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Zinc, an essential element required for cell proliferation and differentiation, plays a role in a diverse array of cellular functions, including acting as a cofactor for numerous enzymes and transcription factors and as a neuroregulator. The zinc transporter (ZnT) family regulates the supply of zinc within cells, and its members are characterized by containing six membrane-spanning domains, a large histidine-rich intracellular loop, and a C-terminal tail. ZnT-1, a ubiquitous protein of approximately 60 kDa, localizes to the plasma membrane to aid in the export of zinc out of cells.
Specifications

Specifications

MED18
Polyclonal
Unconjugated
MED18
MED18, mediator complex subunit 18, p28b
Rabbit
Antigen Affinity Chromatography
RUO
54797, 67219, 682988
-20°C
Liquid
Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9BUE0, Q9CZ82
MED18
MED18 Fusion Protein Ag7598
20 μL
Primary
Mouse, Rat, Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers