missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
MED24 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-74599AF594
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
MED24 Polyclonal antibody specifically detects MED24 in Human, Mouse samples. It is validated for Western Blot
Specifikationer
| MED24 | |
| Polyclonal | |
| Western Blot | |
| cofactor required for Sp1 transcriptional activation, subunit 4, 100kDa, CRSP100, DRIP100CRSP complex subunit 4, hTRAP100, KIAA0130CRSP4, mediator complex subunit 24ARC100, MGC8748, subunit 24 homolog, thyroid hormone receptor associated protein 4, Thyroid hormone receptor-associated protein 4, Thyroid hormone receptor-associated protein complex 100 kDa component, Trap100, TRAP100THRAP4, Vitamin D3 receptor-interacting protein complex 100 kDa component, vitamin D3 receptor-interacting protein complex component DRIP100 | |
| The immunogen recognized by this antibody maps to a region between residue 939 and 989 of human thyroid hormone receptor-associated protein complex 100 kDa component (mediator complex subunit 24 ) using the numbering given in entry NP_055630.2 (GeneID 986). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 9862 | |
| Store at 4C in the dark. | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering