missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED31 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16590-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
MED31 is the component of the Mediator complex, a coactivator involved in the regulated transcription of nearly all RNA polymerase II-dependent genes.Specifications
MED31 | |
Polyclonal | |
Unconjugated | |
MED31 | |
3110004H13Rik, CGI 125, hSOH1, MED31, mediator complex subunit 31, Mediator complex subunit SOH1, Soh1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
287475, 51003, 67279 | |
-20°C | |
Liquid |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9CXU1, Q9Y3C7 | |
Med31 | |
MED31 Fusion Protein Ag9008 | |
20 μL | |
Primary | |
Rat, Human, Mouse | |
Antibody | |
IgG |