missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MED9 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27452-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
In mammalian cells, transcription is regulated in part by high molecular weight co-activating complexes that mediate signals between transcriptional activators and RNA polymerase II (Pol II). The mediator complex is one such multi-protein structure that functions as a bridge between regulatory proteins and Pol II, thereby regulating Pol II-dependent transcription. Med9 (mediator complex subunit 9), also known as MED25, is a 146 amino acid nuclear protein and component of the mediator complex. Med9 directly interacts with Med4 and is encoded by a gene that maps to human chromosome 17p11.2. Chromosome 17 comprises over 2.5% of the human genome, encodes over 1,200 genes and is associated with two key tumor suppressor genes, namely, p53 and BRCA1.Specifications
MED9 | |
Polyclonal | |
Unconjugated | |
MED9 | |
MED25, MED9, mediator complex subunit 9 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55090 | |
-20°C | |
Liquid |
Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9NWA0 | |
MED9 | |
MED9 Fusion Protein Ag26158 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |